Learn More
transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A) (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TCEB3 protein.
Supplier: Abnova Corporation H00006924B01
Description
This gene encodes the protein elongin A, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. [provided by RefSeq
Sequence: MAAESALQVVEKLQARLAANPDPKKLLKYLKKLSTLPITVDILAETGVGKTVNSLRKHEHVGSFARDLVAQWKKLVPVERNAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMYVDHYRSLEEDQEPIVSHQKPGKGHSNAFQDRLGASQERHLGEPHGKGVVSQNKEHKSSHKDKRPVDAKSDEKASVVSREKSHKALSKEENRRPPSGDNAREKPPSSGVKKEKDREGSSLKKKCLPPSEAASDNHLKKPKHRDPEKAKLDKSKQGLDSFDTGKGAGDLLPKVKEKGSNNLKTPEGKVKTNLDRKSLGSLPKVEETDMEDEFEQPTMSFESYLSYDQPRKKKKKIVKTSATALGDKGLKKNDSKSTGKNLDSVQKLPKVNKTKSEKPAGADLAKLRKVPDVLPVLPDLPLPAIQANYRPLPSLELISSFQPKRKAFSSPQEEEEAGFTGRRMNSKMQVYSGSKCAYLPKMMTLHQQCIRVLKNNIDSIFEVGGVPYSVLEPVLERCTPDQLYRIEEYNHVLIEETDQLWKVHCHRDFKEERPEEYESWREMYLRLQDAREQRLRVLTKNIQFAHANKPKGRQAKMAFVNSVAKPPRDVRRRQEKFGTGGAAVPEKIKIKPAPYPMGSSHASASSISFNPSPEEPAYDGPSTSSAHLAPVVSSTVSYDPRKPTVKKIAPMMAKTIKAFKNRFSRRSpecifications
transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TCEB3 protein. | |
TCEB3 | |
FLJ38760/FLJ42849/SIII/TCEB3A | |
Mouse | |
50 μL | |
Transcription Regulation | |
Primary | |
Human | |
Serum |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
NM_003198.1 | |
TCEB3 | |
TCEB3 (NP_003189.1, 1 a.a. ∼ 772 a.a) full-length human protein. | |
RUO | |
Yes | |
6924 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.