Learn More
Description
Specifically detects TCF-4 Clone: 6H5-3 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry
Specifications
Specifications
| Antigen | TCF-4 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6H5-3 |
| Conjugate | Alexa Fluor 488 |
| Formulation | Purified mouse antibody conjugate in PBS with 15mg/ml BSA and 0.1 % sodium azide. |
| Gene Accession No. | Q9NQB0 |
| Gene Symbols | TCF7L2, TCF4 |
| Host Species | Mouse |
| Immunogen | Amino acids 31-331 of human TCF-4. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.