Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TCF-4 Alexa Fluor 647, Clone: 6H5-3,

Catalog No. 05511AF647M Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05-511-AF647M 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-511-AF647M Supplier MilliporeSigma™ Supplier No. 05511AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TCF-4 Clone: 6H5-3 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry

Transcription factor 7-like 2 (UniProt Q9NQB0; also known as HMG box transcription factor 4, hTCF-4, T-cell factor 4, T-cell-specific transcription factor 4, TCF-4) is encoded by the TCF7L2 (also known as TCF4) gene (Gene ID 6934) in human. TCF4, also known as TCF7L2, is expressed widely during development. Gene targeting study indicates that it is required to maintain the crypt stem cells of the small intestine. TCF4 has many different splicing isoforms and they are expressed differentially in tissues and in cancers of different stages. Studies also indicate that variant of the TCF4 gene confers an increased risk of type 2 diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF-4
Applications Immunocytochemistry
Classification Monoclonal
Clone 6H5-3
Conjugate Alexa Fluor 647
Formulation Purified mouse antibody conjugate in PBS with 15mg/ml BSA and 0.1 % sodium azide.
Gene Accession No. Q9NQB0
Gene Symbols TCF7L2, TCF4
Host Species Mouse
Immunogen Amino acids 31-331 of human TCF-4.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) CAB97213.1
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.