Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF-4 Mouse anti-Human, Mouse, Clone: 6H5-3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05511
Description
Use Anti-TCF-4 Antibody, clone 6H5-3 (Mouse Monoclonal Antibody) validated in EMSA, immunocytochemistry, immunohistochemistry, immunoprecipitation, western blotting to detect TCF-4 also known as Immunoglobulin transcription factor 2, SL3-3 enhancer factor 2.
Specifications
| TCF-4 | |
| Monoclonal | |
| Unconjugated | |
| CAB97213.1 | |
| Mouse | |
| Protein G chromatography | |
| Primary | |
| −20°C for up to 12 months. | |
| IgG2a |
| Electromobility Shift Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 6H5-3 | |
| Purified mouse monoclonal IgG2a in buffer containing 0.07MTris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide and 30% glycerol. | |
| TCF4, TCF-4, TCF7L2 | |
| Amino acids 31-331 of human TCF-4. | |
| 200 μg | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction