Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF-7/TCF-1 Mouse, PE, Clone: S33-966, BD

Catalog No. BDB564217 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564217 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564217 Supplier BD Biosciences Supplier No. 564217
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S33-966 monoclonal antibody specifically binds to TCF-7 (Transcription factor 7) which is also known as TCF-1 (T-cell factor 1). TCF-7 is a member of the High Mobility Group (HMG) DNA binding protein family of transcription factors. TCF-7 is expressed in thymocytes, T lymphocytes and proliferating epithelial cells. TCF-7 is expressed in the earliest T cell precursors and serves as a critical transcriptional regulator that is involved in the differentiation of thymocytes to become mature T lymphocytes. TCF-7 is necessary for the survival of immature CD4+CD8+ thymocytes. TCF-7 can also promote the differentiation of mature peripheral T cells to become Th2-like cells. TCF-7 may also function as a transcriptional repressor of certain target genes including CTNNB1 and TCF7L2 . The S33-966 antibody crossreacts with human TCF-7.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF-7/TCF-1
Applications Flow Cytometry
Classification Monoclonal
Clone S33-966
Concentration 0.2mg/mL
Conjugate PE
Description TCF7; Transcription factor 7; TCF-1; TCF1; T-cell factor 1
Host Species Mouse
Immunogen Mouse TCF-7 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.