Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF21 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535055
Description
TCF21 encodes a transcription factor of the basic helix-loop-helix family. The TCF21 product is mesoderm specific, and expressed in embryonic epicardium, mesenchyme-derived tissues of lung, gut, gonad, and both mesenchymal and glomerular epithelial cells in the kidney. Two transcript variants encoding the same protein have been found for this gene.
Specifications
TCF21 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O43680 | |
TCF21 | |
KLH conjugated synthetic peptide between 144-173 amino acids from the C-terminal region of human TCF21 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
TCF21 | |
TCF21; BHLHA23; POD1; Transcription factor 21; Capsulin; Class A basic helix-loop-helix protein 23; Epicardin; Podocyte-expressed 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6943 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction