Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCHP Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526363
Description
This antibody is predicted to react with bovine based on sequence homology.
TCHP may act as a ′capping′ or ′branching′ protein for keratin filaments in the cell periphery. This protein may regulate K8/K18 filament and desmosome organization mainly at the apical or peripheral regions of simple epithelial cells.
Specifications
TCHP | |
Polyclonal | |
TCHP | |
TpMs, mitochondrial protein with oncostatic activity, mitostatin, trichoplein keratin filament-binding protein, tumor suppressor protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84260 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9BT92 | |
TCHP | |
KLH conjugated synthetic peptide between 291-320 amino acids from the central region of human TCHP | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction