Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCL1 Mouse anti-Human, MilliporeSigma™

Catalog No. 05954MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-954-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-954-MI Supplier MilliporeSigma Supplier No. 05954
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-TCL1 Antibody is validated for use in western blot, immunohistochemistry for the detection of TCL1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCL1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Gene Accession No. NM_021966
Gene Symbols TCL1A, TCL1, TCL1-PEN
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 60-73 (RPMTPTQIGPSLLP) of human TCL1A
Purification Method Cell culture supernatant
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.