Learn More
transcobalamin I (vitamin B12 binding protein, R binder family), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the vitamin B12-binding protein family. This family of proteins, alternatively referred to as R binders, is expressed in various tissues and secretions. This protein is a major constituent of secondary granules in neutrophils and facilitates the transport of cobalamin into cells. [provided by RefSeq
Specifications
Specifications
| Antigen | transcobalamin I (vitamin B12 binding protein, R binder family) |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human TCN1 protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TCN1 |
| Gene Accession No. | NM_001062.2 |
| Gene Alias | TC1/TCI |
| Gene Symbols | TCN1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.