Learn More
TCR ab Mouse anti-Human, FITC, Clone: WT31, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 347773
Description
Clone WT31 is derived from hybridization of mouse Sp2/0-Ag14 myeloma cells with spleen cells from BALB/c mice immunized with human thymocytes.
WT31 (Anti–TCR-α/β-1) recognizes a conformational epitope formed by the T-cell receptor (TCR) for antigen and the CD3 epsilon chain. The α/β TCR is a disulfide-linked 80-kdalton (kDa) heterodimer consisting of a 44-kDa α chain and a 37-kDa β chain.

Specifications
TCR ab | |
Monoclonal | |
50μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human thymocytes | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. | |
IgG1 κ |
Flow Cytometry | |
WT31 | |
FITC | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.