Learn More
Description
Host Species: Hamster
Clone H57-597
Isotype IgG2, λ1
Species Reactivity Ms
Immunogen TCR affinity-purified from mouse T-cell hybridoma DO-11.10
Flow Cytometry
Specifications
Specifications
| Antigen | TCR Beta Chain |
| Classification | Monoclonal |
| Clone | H57-597 |
| Concentration | 0.2mg/mL |
| Host Species | Hamster |
| Immunogen | TCR affinity-purified from mouse T-cell hybridoma DO-11.10 |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.