Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR β chain Hamster anti-Mouse, FITC, Clone: H57-597, BD

Catalog No. BDB553170 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553170 0.1 mg
BDB553171 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553170 Supplier BD Biosciences Supplier No. 553170
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The H57-597 antibody reacts with a common epitope of the β; chain of the T-cell Receptor (TCR) complex on α;β; TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β; -chain may form homodimers or pair with the pre-TCR α; -chain on the surface of immature thymocytes before expression of the TCR α; -chain. Plate-bound or soluble H57-597 antibody activates α;β; TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR β chain
Applications Flow Cytometry
Classification Monoclonal
Clone H57-597
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen TCR affinity-purified from mouse T-cell hybridoma DO-11.10
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.