Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCRγ/δ Mouse anti-Human, Unlabeled, Clone: B1, BD

Catalog No. BDB555715 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555715 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555715 Supplier BD Biosciences Supplier No. 555715
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recognizes the γ/δ T-cell receptor (TCR). This receptor complex consists of two disulfide-linked glycoproteins, a γ chain (45-60kDa) and a δ subunit (40-60kDa). γ/δ TCR is expressed in less than 10% of human peripheral T cells. The physiological significance of γ/δ + T cells is still unknown. There is evidence indicating that these cells recognize bacterial ligands and some tumor cells. Reports suggest that γ/δ + T cells may play a role in the immune reaction during infection and in regulation of pathphysiological autoimmune responses. Due to the fact that CD3 and TCRγδ antibodies may recognize epitopes which are very close to each other and that CD3 was interfering with the clone B1 antibody binding but not with the clone 11F2 antibody, we recommend to use clone 11F2 antibody to perform double staining TCRγδ and CD3. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: B1
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCRγ/δ
Applications Flow Cytometry
Classification Monoclonal
Clone B1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.