Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR γδ Hamster anti-Mouse, Unlabeled, Clone: GL4, BD

Catalog No. BDB553180 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553180 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553180 Supplier BD Biosciences Supplier No. 553180

Hamster Monoclonal Antibody

The GL4 antibody reacts with a common epitope of the δ chain of the γδ T-cell Receptor (TCR) complex on γδ TCR-expressing T lymphocytes and NK-T cells of all mouse strains tested. It does not react with αβ TCR-bear

Host Species: Hamster
Clone: GL4
Isotype: IgG2 κ
Species Reactivity: Mouse
Immunogen: C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes

Flow Cytometry, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR γδ
Applications Flow Cytometry
Classification Monoclonal
Clone GL4
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Bovine, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.