Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR γδ Hamster anti-Mouse, Unlabeled, Clone: UC7-13D5, BD

Catalog No. BDB553182 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553182 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553182 Supplier BD Biosciences Supplier No. 553182

Hamster Monoclonal Antibody

The UC7-13D5 antibody reacts with the γδ T-cell Receptor (TCR) complex on γδ TCR-expressing T lymphocytes and NK-T cells of all mouse strains tested. It does not react with the αβ TCR-bearing T cells. In the mouse, cellexpressing the γδ TCR are found in the thymus, intestinal epithelium, epidermis, dermis, pulmonary epithelium, peritoneum, liver, and peripheral lymphoid organs. Plate-bound UC7-13D5 antibody activates γδ TCR-bearing T cells, and in vivo administration of the mAb depletes peripheral γδ TCR-bearing T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR γδ
Applications Flow Cytometry
Classification Monoclonal
Clone UC7-13D5
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Hamster
Immunogen Not Reported
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Bovine, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG κ3
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.