Learn More
TCR gammaδ Hamster anti-Mouse, FITC, Clone: GL3, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 561996
Description
The GL3 monoclonal antibody specifically binds to a common epitope of the δ chain of the T-cell Receptor (TCR) complex on γδ TCR-expressing T lymphocytes and NK-T cells of all mouse strains tested. It does not react with αβ TCR-bearing T cells. In the mouse, cells expressing the γδ TCR are found in the thymus, intestinal epithelium, epidermis, dermis, pulmonsry epithelium, peritoneum, liver, and peripheral lymphoid organs.

Specifications
TCR gammaδ | |
Monoclonal | |
0.05mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes | |
0.05 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
GL3 | |
FITC | |
Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.