Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR αβ Mouse anti-Human, FITC, Clone: T10B9.1A-31, BD

Catalog No. BDB555547 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555547 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555547 Supplier BD Biosciences Supplier No. 555547
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with a monomorphic determinant of αβ T-cell receptor expressed on greater than 95% of normal peripheral blood CD3+ T cells. The αβ TCR recognizes a peptide bound to MHC leading to T-cell activation. The T10B9.1A-31 antibody induces T-cell activation in the immobilized form and is useful in flow cytometry for studying αβ+ T-cell populations. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR αβ
Applications Flow Cytometry
Classification Monoclonal
Clone T10B9.1A-31
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Peripheral Blood T Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.