Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR αβ Mouse anti-Human, PE, Clone: T10B9.1A-31 (also known as T10B9), BD

Catalog No. BDB561674 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561674 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561674 Supplier BD Biosciences Supplier No. 561674
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The T10B9.1A-31 monoclonal antibody specifically binds to a monomorphic determinant on the αβ T-cell receptor expressed on greater than 95% of normal peripheral blood CD3+ T cells. The αβ TCR recognizes a peptide bound to MHC leading to T-cell activation. The T10B9.1A-31 antibody induces T-cell activation in the immobilized form and is useful in flow cytometry for studying αβ+ T-cell populations.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR αβ
Applications Flow Cytometry
Classification Monoclonal
Clone T10B9.1A-31 (also known as T10B9)
Conjugate PE
Description αβ TCR; TCR alpha-beta
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Peripheral Blood T Cells
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.