Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCRγδ Mouse anti-Human, PerCP-Cy5.5, Clone: B1, BD

Catalog No. BDB564157 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564157 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564157 Supplier BD Biosciences Supplier No. 564157
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The B1 monoclonal antibody specifically binds to the γδ T cell receptor ( γδ TCR). This receptor complex consists of two disulfide-linked transmembrane glycoproteins, a γchain (45-60 kDa) and a δ subunit (40-60 kDa). The γδ TCR is associated with the signal-transducing CD3 complex. The γδ TCR is expressed by thymocytes and by peripheral T cell subsets (γδ T cells) that are located in the blood, liver, skin and various lymphoid and mucosal tissues. γδ T cells contribute to both innate and adaptive immune responses to infections and tumors. Reports suggest that γδ

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCRγδ
Applications Flow Cytometry
Classification Monoclonal
Clone B1
Concentration 0.2mg/mL
Conjugate PerCP-Cyanine5.5
Description TCRgd; γδ TCR; TRG@, TRD@; TCRG, TCRD; TCR gamma delta
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.