Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCRαβ Mouse anti-Human, PE, Clone: IP26 (also known as IP26A), BD

Catalog No. BDB564728 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564728 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564728 Supplier BD Biosciences Supplier No. 564728
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The IP26 monoclonal antibody specifically recognizes a m onomorphic determinant on the αβ T-cell receptor (TCR αβ ). The TCR αβ is a disulfide-linked 80 kDa heterodimer consisting of a 44 kDa α chain and a 37 kDa β chain. The TCR αβ is normally expressed on 50-70% of thymocytes and on a large fraction of mature T cells including greater than 95% of peripheral blood CD3+ T cells. The TCRαβ serves as a receptor for peptide antigens bound to MHC molecules. The IP26 antibody is mitogenic and useful for the flow cytometric analysis of TCR αβ+ T-cell subsets.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCRαβ
Applications Flow Cytometry
Classification Monoclonal
Clone IP26 (also known as IP26A)
Conjugate PE
Description alpha-beta T cell receptor, TCRAB, TRA@/TRB@; IP26A
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human T Cell Clone
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.