Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR αβ Mouse anti-Human, PE, Clone: T10B9.1A-31, BD

Catalog No. BDB555548 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555548 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555548 Supplier BD Biosciences Supplier No. 555548
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

β2-microglobulin is the 12kDaβ chain which noncovalently associates with the 44kDa α chain to form the HLA Class I antigen complex. This antibody reacts with free or complexed β2-microglobulin. T_99 does not react with β2-microglobulin of other species. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections using TUF pretreatment. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR αβ
Applications Flow Cytometry
Classification Monoclonal
Clone T10B9.1A-31
Conjugate PE
Description αβ TCR; TCR alpha-beta
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Peripheral Blood T Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.