Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tdp43 Mouse anti-Human, Mouse, Rat, Clone: 3H8, MilliporeSigma™
GSA_VA

Catalog No. MABN45MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN45MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN45MI Supplier MilliporeSigma Supplier No. MABN45
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect TDP using this mouse monoclonal antibody, Anti-Tdp43 Antibody, clone 3H8 validated for use in western blotting and immunohistochemistry.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tdp43
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3H8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS with 0.05% sodium azide with 10% glycerol.
Gene Accession No. NP_031401
Gene Symbols TARDBP, TDP43
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to the N-terminus of human Tdp43.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.