Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146821
Description
TDRD3 Polyclonal specifically detects TDRD3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
TDRD3 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
TDRD3 | |
The immunogen for this product maps to a region between residue 601 and 651 of human tudor domain containing 3 using the numbering given in entry NP_110421.1 (GeneID 81550). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin | |
FLJ21007, tudor domain containing 3, tudor domain-containing protein 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
81550 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction