Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEF-1 Mouse, Unlabeled, Clone: 31, BD

Catalog No. BDB610922 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610922 50 μg
BDB610923 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610922 Supplier BD Biosciences Supplier No. 610922
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TEF-1 (transcription enhancer factor-1) is a member of a family of transcription factors that contain an evolutionarily conserved DNA binding domain (DBD), TEA/ATTS. TEF-1 was identified as a HeLa cell transcriptional activator that bound, via its TEA domain, to the GT-IIC and Sph enhancer elements. Comparison of various enhancers indicates that TEF-1 binds to highly degenerate DNA sequences and cooperatively interacts with tandem repeats of its binding sites, which increases its enhancer activity. However, the DNA binding activities of TEF-1 are modulated by sequences outside the DBD. The N-terminal basic portion of TEF-1 contains the DBD. The C-terminal portion contains the transactivation domain and a zinc finger-like motif that is thought to be involved in protein dimerization or interaction with additional transcription factors. TEF-1 induced transcription requires a limiting transcriptional intermediary factor (TIF) and a TATA-binding protein associated factor (hTAFIIs). Although TEF-1 is expressed ubiquitously during embryogenesis, its expression is limited to adult kidney, heart, brain, skeletal muscle, and lungs. Disruption of murine TEF-1 gene results in fetal death.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TEF-1
Applications Western Blot
Classification Monoclonal
Clone 31
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human TEF-1 aa. 86-199
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.