Learn More
Tenascin Rabbit anti-Human, Chicken, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB19013
Description
Anti-Tenascin Antibody is an antibody against Tenascin for use in immunohistochemistry, immunohistochemistry (paraffin).
Specifications
Tenascin | |
Polyclonal | |
Affinity Purified using immobilized chicken tenascin. Provided as a liquid in PBS, containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Chicken, Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
TNC; Hexabrachion; HXB; GMEM; TN; Neuronectin; Tenascin-C; MGC167029; JI; TN-C; Cytotactin | |
Purified tenascin from embryonic chicken brain. | |
50 μg | |
NM_002160.2 | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. |