Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB19011MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB19011MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB19011MI Supplier MilliporeSigma Supplier No. AB19011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Tenascin Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Tenascin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tenascin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified using immobilized human tenascin; presented as a liquid in PBS containing no preservatives.
Gene Symbols TNC; Hexabrachion; HXB; GMEM; TN; Neuronectin; Tenascin-C; MGC167029; JI; TN-C; Cytotactin
Host Species Rabbit
Immunogen Purified human tenascin from glioma culture supernatant.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002160.2
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.