Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin Mouse anti-Guinea Pig, Human, Rabbit, Clone: DB7, MilliporeSigma™

Catalog No. MAB19101MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB19101MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB19101MI Supplier MilliporeSigma Supplier No. MAB19101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Tenascin using this Anti-Tenascin Antibody, clone DB7 validated for use in immunofluoresence, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tenascin
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone DB7
Conjugate Unconjugated
Formulation The purified antibody is in PBS solution containing 1.0% (w/v) BSA and 0.1% (w/v) sodium azide.
Gene Symbols ITGAM, MO1A, CD11b, CR3A, MAC1A, CD11B, MGC117044, MAC-1
Host Species Mouse
Immunogen Tenascin polypeptides isolated from spent culture supernatant of human fibroblasts isolated by affinity chromatography.
Quantity 50 μg
Primary or Secondary Primary
Target Species Guinea Pig, Human, Rabbit
Content And Storage 2-8°C.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.