Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TER-119, APC, Clone: TER-119,

Catalog No. MABF1431MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1431MI 100 μg
MABF1430MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1431MI Supplier MilliporeSigma™ Supplier No. MABF1431
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects TER-119 Clone: TER-119 in Mouse samples, and it is validated for Flow Cytometry

The TER-119 antibody is named for the antigen to which it binds, a 52kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TER-119
Applications Flow Cytometry
Classification Monoclonal
Clone TER-119
Conjugate APC
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. Q9ES52
Gene Symbols Inpp5d, 7a33, Ship, Ship1
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Purified by size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.