Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TER-119, FITC, Clone: TER-119,

Rat Monoclonal Antibody

Supplier:  MilliporeSigma™ MABF1434

 View more versions of this product

Catalog No. MABF1434MI


Only null left
Add to Cart

Description

Description

Specifically detects TER-119 Clone: TER-119 in Mouse samples, and it is validated for Flow Cytometry

The TER-119 antibody is named for the antigen to which it binds, a 52kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TER-119
Monoclonal
FITC
Q9ES52
Rat
Purified by size-exclusion chromatography
RUO
Primary
2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Flow Cytometry
TER-119
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Inpp5d, 7a33, Ship, Ship1
Proprietary immunogen
25 μg
Inflammation, Immunology
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only