Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terminal Transferase Mouse anti-Human, PE, Clone: E17-1519, BD

Catalog No. BDB347195 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347195 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347195 Supplier BD Biosciences Supplier No. 347195
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-terminal-deoxynucleotidyl transferase (TdT) recognizes a 60kDa polymerase, a nuclear enzyme that catalyzes the template-independent addition of nucleotides to single-stranded DNA primers. It has been reported that TdT is involved in the regulation or translocation or both of DNA and gene rearrangement during normal T- and B-cell development.TdT is present in nuclei of immature T and B lymphocytes found in normal thymus and bone marrow. Some neoplastic cells of acute lymphoblastic lymphoma/leukemia have been shown to have high TdT activity. Mature lymphocytes from normal peripheral blood usually do not contain the enzyme.

Applications:

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Terminal Transferase
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone E17-1519
Concentration 6.0μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Purified Human TdT
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.