Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terminal Transferase Set Mouse anti-Human, PE, Clone: E17-1519, BD

Catalog No. BDB550757 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550757 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550757 Supplier BD Biosciences Supplier No. 550757
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with terminal deoxyribonucleotidyltransferase (TdT), a template-independent DNA polymerase that adds nucleotides to single stranded DNA primers. Western blot analysis of TdT reveals bands of 55, 40, and 15 kDa. TdT is found in normal bone marrow lymphoid progenitor cells and immature thymic lymphocytes. It has been reported that TdT is involved in the regulation and/or translocation of DNA and gene rearrangement during normal T and B cell development.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Terminal Transferase Set
Applications Flow Cytometry
Classification Monoclonal
Clone E17-1519
Conjugate PE
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Includes PE Mouse Anti-Human Terminal transferase (Tdt) (100 tests), PE Mouse IgG1, κ Isotype Control (100 tests)
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.