Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET2 Mouse anti-Human, Mouse, Clone: hT2H21F11, MilliporeSigma™

Catalog No. MABE462MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE462MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE462MI Supplier MilliporeSigma Supplier No. MABE462
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Tet2 using this mouse monoclonal antibody, Anti-TET2 Antibody, clone hT2H 21F11 validated for use in Chromatin immunoprecipitation (ChIP), western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TET2
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone hT2H21F11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001120680
Gene Symbols TET2
Host Species Mouse
Immunogen Recombinant protein corresponding to the N-terminus of human TET2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.