Learn More
Description
This gene encodes a glycoprotein with an approximate molecular weight of 76.5kDa. It is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds one ion of ferric iron. The function of this protein is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. This protein may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter and allergens from serum. [provided by RefSeq
Specifications
Specifications
| Antigen | TF |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 1C2 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant TF. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TF |
| Gene Accession No. | BC059367 |
| Gene Alias | DKFZp781D0156/PRO1557/PRO2086 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.