Learn More
TFAP2A, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human TFAP2A peptide using ARM Technology.
Supplier: Abnova Corporation H00007020K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
AP2-alpha is a 52kDa retinoic acid-inducible and developmentally regulated activator of transcription that binds to a consensus DNA-binding sequence CCCCAGGC in the SV40 and metallothionein (MIM 156350) promoters.[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
TFAP2A | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
TFAP2A | |
TFAP2A | |
A synthetic peptide of human TFAP2A | |
RUO | |
7020 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha) | |
TFAP2A | |
AP-2, AP-2alpha, AP2TF, BOFS, TFAP2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.