Learn More
transcription factor AP-2 beta (activating enhancer binding protein 2 beta), Mouse, Clone: 2F6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TFAP2B.
Supplier: Abnova Corporation H00007021M02
Description
This gene encodes a member of the AP-2 family of transcription factors. AP-2 proteins form homo- or hetero-dimers with other AP-2 family members and bind specific DNA sequences. They are thought to stimulate cell proliferation and suppress terminal differentiation of specific cell types during embryonic development. Specific AP-2 family members differ in their expression patterns and binding affinity for different promoters. This protein functions as both a transcriptional activator and repressor. Mutations in this gene result in autosomal dominant Char syndrome, suggesting that this gene functions in the differentiation of neural crest cell derivatives. [provided by RefSeq
Sequence: DPYSHVNDPYSLNPLHQPQQHPWGQRQRQEVGSEAGSLLPQPRAALPQLSGLDPRRDYHSVRRPDVLLHSAHHGLDAGMGDSLSLHGLGHPGMEDVQSVEDANNSGMNLLSpecifications
transcription factor AP-2 beta (activating enhancer binding protein 2 beta) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003221 | |
TFAP2B | |
TFAP2B (NP_003212, 73 a.a. ∼ 182 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Neurobiology | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Western Blot | |
2F6 | |
Mouse monoclonal antibody raised against a partial recombinant TFAP2B. | |
TFAP2B | |
AP-2B/AP2-B/MGC21381 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
7021 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.