Learn More
transcription factor B1, mitochondrial, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The transcription of genes from mitochondrial DNA requires a mitochondrial RNA polymerase (see POLRMT, MIM 601778) and a DNA-binding transcription factor (see TFAM, MIM 600438). Transcription factor B1 (TFB1M) is a part of this transcription complex.[supplied by OMIM
Specifications
Specifications
| Antigen | transcription factor B1, mitochondrial |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human TFB1M protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TFB1M |
| Gene Accession No. | BC017788.1 |
| Gene Alias | CGI-75/CGI75/mtTFB/mtTFB1 |
| Gene Symbols | TFB1M |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.