Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transcription factor B2, mitochondrial, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004707 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-707 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-707 Supplier Abnova Corporation Supplier No. H00064216A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TFB2M.

Sequence: YLIQMIPRQNLFTKNLTPMNYNIFFHLLKHCFGRRSATVIDHLRSLTPLDARDILMQIGKQEDEKVVNMHPQDFKTLFETIERSKDCAYKWLYDETLEDR

Specifications

Antigen transcription factor B2, mitochondrial
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TFB2M.
Formulation 50% glycerol
Gene TFB2M
Gene Accession No. NM_022366
Gene Alias FLJ22661/FLJ23182/Hkp1
Gene Symbols TFB2M
Host Species Mouse
Immunogen TFB2M (NP_071761, 297 a.a. ∼ 396 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 64216
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.