Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFE3 Mouse anti-Mouse, Unlabeled, Clone: G138-312, BD

Catalog No. BDB554263 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554263 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554263 Supplier BD Biosciences Supplier No. 554263
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TFE3 is a ubiquitously expressed 50kDa transcription factor containing both basic helix-loop-helix (bHLH) and leucine zipper (ZIP) motifs. It was initially identified as a protein which bound to the μE3 site in the immunoglobulin heavy chain enhancer. In addition, it binds to the MLTF/USF site in the adenovirus major late promoter. Upon binding as a dimer, it induces a minor groove-oriented bend in the DNA, similar to other HLH proteins such as TFEB, USF, myc, and max. Binding results in transcriptional activation and is mediated through the basic region located just N-terminal to the HLH domain. Clone G138-312 recognizes TFE3. The antibody was raised against a bacterially expressed TFE3-L protein consisting of 327 amino acids encoded by nucleotides 361-2469 of the cloned cDNA.

Host Species: Mouse
Clone: G138-312
Isotype: IgG1
Species Reactivity: Human
Immunogen: TFE3-L Protein
Formula Weight [Chemical]: 50kDa

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TFE3
Applications Western Blot
Classification Monoclonal
Clone G138-312
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen TFE3-L Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.