Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trefoil factor 3 (intestinal), Mouse, Clone: 3G11, Abnova™

Catalog No. 89001013 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-013 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-013 Supplier Abnova Corporation Supplier No. H00007033M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TFF3.

Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer and affect healing of the epithelium. This gene is expressed in goblet cells of the intestines and colon. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21. [provided by RefSeq

Sequence: EEYVGLSANQCAVPAKDRVDCGYPHVTPKECNNRGCCFDSRIPGVPWCFKPLQEAECTF

Specifications

Antigen trefoil factor 3 (intestinal)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant TFF3.
Formulation PBS with no preservative; pH 7.4
Gene TFF3
Gene Accession No. BC017859
Gene Alias HITF/ITF/TFI/hP1.B
Gene Symbols TFF3
Host Species Mouse
Immunogen TFF3 (AAH17859.1, 15 a.a. ∼ 73 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7033
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.