Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFIIB Mouse, Unlabeled, Clone: 24, BD

Catalog No. BDB610462 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610462 50 μg
BDB610463 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610462 Supplier BD Biosciences Supplier No. 610462

Mouse Monoclonal Antibody

TFIIB is a ubiquitous factor required for transcription initiation by RNA polymerase II. Studies suggest that TFIIB serves as a bridge between the “TATA”-binding factor (TFIID) and RNA polymerase II during pre-initiation complex assembly and that TFIIB can be a target of acidic activators. An essential component of the machinery that transcribes protein-coding genes, the three-dimensional structure of the human TFIIB appears to consist of two direct repeats that adopt similar alpha-helical folds, conferring pseudo-twofold symmetry. An extensive, central basic surface, including an amphipathic alpha helix, is critical to the function of TFIIB as a bridge between the TBP-promoter complex and transcription factors. TFIIB has a calculated molecular weight of 33kDa and has been reported to be observed to migrate between 33-40kDa.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TFIIB
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 24
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Frog, Human, Mouse, Rat
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.