Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-β1 Rat anti-Human, Pig, Unlabeled, Clone: A75-2, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 555052
Description
The A75-2 antibody reacts with mouse, pig, human TGF-β1. The immunogen used to generate this hybridoma was recombinant mouse TGF-β1.
ELISA, Western Blotting

Specifications
TGF-β1 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Mouse TGF-β1 | |
0.5 mg | |
Immunology | |
Human, Mouse, Pig | |
IgG2a κ |
ELISA | |
A75-2 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction