Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-Beta 1 Mouse anti-Human, Brilliant Violet 421, Clone: TW4-9E7, BD

Catalog No. BDB562962 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562962 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562962 Supplier BD Biosciences Supplier No. 562962
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TW4-9E7 monoclonal antibody specifically binds to Human Transforming Growth Factor beta-1 (TGF-β1). TGF-β1 is a potent multifunctional cytokine that positively or negatively regulates numerous processes including development, hematopoiesis, tissue remodeling, wound repair, innate and adaptive immunity as well as cancer and autoimmune diseases. TGF-β1 is formed by the enzymatic cleavage of the TGF-β1 propeptide that is encoded by the TGFB1 gene and comprised of the Latency Associated Peptide (LAP) and TGF-β1. Prior to secretion, the dimeric LAP-TGF-β1 propeptide is cleaved resulting in a biologically inactive form of dimeric TGF-β1 that is noncovalently associated with dimeric LAP (latent TGF-β 1). This complex may be expressed on the surface of TGF-β1-producing cells or be further processed by proteolytic removal of LAP to release the biologically active mature form of the soluble TGF-β1 homodimer. Many different cell types synthesize TGF-β1 and express specific receptors for it. The TW4-9E7 antibody recognizes both the intracellular latent bound form of TGF-β1 along with the membrane bound form of TGF-β1.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF-Beta 1
Applications Flow Cytometry
Classification Monoclonal
Clone TW4-9E7
Conjugate Brilliant Violet 421
Description TGFB1; TGFbeta1; TGF-beta-1; Transforming growth factor, beta 1; CED; LAP
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human TGF-β1 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.