Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGF-beta RII Fluorescein-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
$186.20 - $397.00
Specifications
Antigen | TGF-beta RII |
---|---|
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | Fluorescein |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB2411F025
![]() |
R&D Systems
FAB2411F025 |
25 Tests |
Each for $186.20
|
|
|||||
FAB2411F100
![]() |
R&D Systems
FAB2411F100 |
100 Tests |
Each for $397.00
|
|
|||||
Description
TGF-beta RII Polyclonal specifically detects TGF-beta RII in Human samples. It is validated for Flow Cytometry.Specifications
TGF-beta RII | |
Flow Cytometry | |
Fluorescein | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
TGFBR2 | |
IgG | |
Affinity Purified | |
Detects human TGF-beta RII in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) TGF-beta RII and less than 1% cross-reactivity with recombinant human TGF-beta RIII and rmTGF-beta RI is observed. |
Flow Cytometry 10 uL/10^6 cells | |
Polyclonal | |
Goat | |
Human | |
P37173.2 | |
7048 | |
Mouse myeloma cell line NS0-derived recombinant human TGF-beta RII Ile24-Asp159 Accession # P37173.2 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title