Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGF-beta RIII Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB242
This item is not returnable.
View return policy
Description
TGF-beta RIII Monoclonal specifically detects TGF-beta RIII in Human samples. It is validated for Western Blot.Specifications
| TGF-beta RIII | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| betaglycan, betaglycan proteoglycan, BGCAN, TBRIII, TGF-beta receptor type 3, TGF-beta receptor type III, TGFbetaRIII, TGFBR3, TGF-bRIII, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7049 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 31349 | |
| Western Blot 1 ug/mL | |
| AAA67061 | |
| TGFBR3 | |
| Mouse myeloma cell line NS0-derived recombinant human TGF-beta RIII Gly21-Asp781 Accession # AAA67061 | |
| 500 μg | |
| Primary | |
| Detects human TGF-beta RIII in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant rat MIS RII, recombinant human (rh) TGF-beta RI, rhTGF-beta RII, recombinant mouse TGF-beta RII, or rhTGF-beta RIIb. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction