Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TGF-beta RII Antibody, R&D Systems™

Mouse Monoclonal Antibody
$227.50 - $2070.00
Specifications
Antigen | TGF-beta RII |
---|---|
Clone | 176510 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2411SP
![]() |
R&D Systems
MAB2411SP |
25 μg |
Each for $227.50
|
|
|||||
MAB2411100
![]() |
R&D Systems
MAB2411100 |
100 μg |
Each for $561.50
|
|
|||||
MAB2411500
![]() |
R&D Systems
MAB2411500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
TGF-beta RII Monoclonal specifically detects TGF-beta RII in Human samples. It is validated for Western Blot.Specifications
TGF-beta RII | |
Western Blot 1 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
NP_001020018 | |
7048 | |
Mouse myeloma cell line NS0-derived recombinant human TGF-beta RII isoform 2 Thr23-Asp184 Accession # NP_001020018 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TGF-beta RII and TGF-beta RIIb in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human TGF-beta RIII, recombinant mouse (rm) TGF-beta RII, or rmTGF-beta RI. |
176510 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
TGFBR2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title