Learn More
transforming growth factor, beta 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | transforming growth factor, beta 2 |
| Applications | In situ PLA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human TGFB2 protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TGFB2 |
| Gene Accession No. | NM_003238 |
| Gene Alias | MGC116892/TGF-beta2 |
| Gene Symbols | TGFB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.