Learn More
transforming growth factor, beta-induced, 68kDa, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy. [provided by RefSeq
Specifications
Specifications
| Antigen | transforming growth factor, beta-induced, 68kDa |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant TGFBI. |
| Formulation | 50% glycerol |
| Gene | TGFBI |
| Gene Accession No. | BC000097 |
| Gene Alias | BIGH3/CDB1/CDG2/CDGG1/CSD/CSD1/CSD2/CSD3/EBMD/LCD1 |
| Gene Symbols | TGFBI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.