Learn More
transforming growth factor, beta receptor II (70/80kDa), Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the Ser/Thr protein kinase family and the TGFB receptor subfamily. The encoded protein is a transmembrane protein that has a protein kinase domain, forms a heterodimeric complex with another receptor protein, and binds TGF-beta. This receptor/ligand complex phosphorylates proteins, which then enter the nucleus and regulate the transcription of a subset of genes related to cell proliferation. Mutations in this gene have been associated with Marfan Syndrome, Loeys-Deitz Aortic Aneurysm Syndrome, and the development of various types of tumors. Alternatively spliced transcript variants encoding different isoforms have been characterized. [provided by RefSeq
Specifications
Specifications
| Antigen | transforming growth factor, beta receptor II (70/80kDa) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant TGFBR2. |
| Formulation | 50% glycerol |
| Gene | TGFBR2 |
| Gene Accession No. | NM_001024847 |
| Gene Alias | AAT3/FAA3/LDS1B/LDS2B/MFS2/RIIC/TAAD2/TGFR-2/TGFbeta-RII |
| Gene Symbols | TGFBR2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.