Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBR3 Mouse anti-Human, Mouse, Clone: 1C5H11, MilliporeSigma™

Catalog No. MABC710MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC710MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC710MI Supplier MilliporeSigma Supplier No. MABC710
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TGFBR3, clone 1C5H11 Antibody is an antibody against TGFBR3 for use in western blotting, flow cytometry and ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGFBR3
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1C5H11
Conjugate Unconjugated
Formulation Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Gene Symbols TGFBR3
Host Species Mouse
Immunogen Purified recombinant fragment of human TGFBR3 expressed in E. Coli
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.