Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transglutaminase 7, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004771 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-771 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-771 Supplier Abnova Corporation Supplier No. H00116179A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TGM7.

Transglutaminases (TGM; EC 2.3.2.13) are a family of structurally and functionally related enzymes that stabilize protein assemblies through the formation of gamma-glutamyl-epsilon lysine crosslinks. For additional background information on transglutaminases, see TGM1 (MIM 190195).[supplied by OMIM

Sequence: MLGPQRASLPFLDLLESGGLRDQPAQLQLHLARIPEWGQDLQLLLRIQRVPDSTHPRGPIGLVVRFCAQALLHGGGTQKPFWRHTVRMNLDFGKETQWPL

Specifications

Antigen transglutaminase 7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TGM7.
Formulation 50% glycerol
Gene TGM7
Gene Accession No. NM_052955
Gene Alias TGMZ
Gene Symbols TGM7
Host Species Mouse
Immunogen TGM7 (NP_443187, 466 a.a. ∼ 565 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 116179
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.